Anti-CPEB3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Leverantör: United States Biological
USBI034162-PEEA 7175 SEK
USBI034162-PE
Anti-CPEB3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: CPEB3 (cytoplasmic polyadenylation element binding protein 3)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human