Anti-CPEB4 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Leverantör: United States Biological
USBI034163-PEEA 9900 SEK
USBI034163-PE
Anti-CPEB4 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!
Type: Primary
Antigen: CPEB4 (cytoplasmic polyadenylation element binding protein 4)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse