Anti-CPNE3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Leverantör: United States Biological
USBI034169-APEA 9900 SEK
USBI034169-AP
Anti-CPNE3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: CPNE3 (copine III)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human