Anti-CPNE5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverantör: United States Biological
USBI034170-FITCEA 9900 SEK
USBI034170-FITC
Anti-CPNE5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: CPNE5 (copine V)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR