Anti-CPNE6 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Leverantör: United States Biological
USBI034171-APCEA 9900 SEK
USBI034171-APC
Anti-CPNE6 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: CPNE6 (copine VI (neuronal))
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse