Anti-CPPED1 Rabbit Polyclonal Antibody (Biotin)

Leverantör: United States Biological
USBI034176-BIOTINEA 9900 SEK
USBI034176-BIOTIN
Anti-CPPED1 Rabbit Polyclonal Antibody (Biotin)
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: CPPED1 (calcineurin-like phosphoesterase domain containing 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR