Anti-CRADD Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Leverantör: US Biological
USBI034199-PEEA 9900 SEK
USBI034199-PE
Anti-CRADD Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: CRADD (CASP2 and RIPK1 domain containing adaptor with death domain)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human