Anti-CREB3L4 Rabbit Polyclonal Antibody

Leverantör: US Biological
USBI034207EA 6825 SEK
USBI034207
Anti-CREB3L4 Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: CREB3L4 (cAMP responsive element binding protein 3-like 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR