Anti-CRELD2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Leverantör: US Biological
USBI034215-PEEA 9900 SEK
USBI034215-PE
Anti-CRELD2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: CRELD2 (cysteine-rich with EGF-like domains 2)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human