Anti-CRIP1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverantör: US Biological
USBI034224-HRPEA 9900 SEK
USBI034224-HRP
Anti-CRIP1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50

Type: Primary
Antigen: CRIP1 (cysteine-rich protein 1 (intestinal))
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR