Anti-CRISP1 Rabbit Polyclonal Antibody

Leverantör: US Biological
USBI034229EA 6825 SEK
USBI034229
Anti-CRISP1 Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: CRISP1 (cysteine-rich secretory protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR