Anti-CRKL Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Leverantör: US Biological
USBI034231-PEEA 9900 SEK
USBI034231-PE
Anti-CRKL Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: CRKL (v-crk avian sarcoma virus CT10 oncogene homolog-like)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse