Anti-CRTRT1 Rabbit Polyclonal Antibody (Biotin)
Leverantör: US Biological
USBI034248-BIOTINEA 9900 SEK
USBI034248-BIOTIN
Anti-CRTRT1 Rabbit Polyclonal Antibody (Biotin)
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: CRTRT1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human