Anti-CRYGB Rabbit Polyclonal Antibody (Biotin)
Leverantör: US Biological
USBI034257-BIOTINEA 9900 SEK
USBI034257-BIOTIN
Anti-CRYGB Rabbit Polyclonal Antibody (Biotin)
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: CRYGB (crystallin, gamma B)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human