Anti-CRYZL1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Leverantör: US Biological
USBI034262-PEEA 9900 SEK
USBI034262-PE
Anti-CRYZL1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: CRYZL1 (crystallin, zeta (quinone reductase)-like 1)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human