Anti-CSDE1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverantör: US Biological
USBI034267-APEA 9900 SEK
USBI034267-AP
Anti-CSDE1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: CSDE1 (cold shock domain containing E1, RNA-binding)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR