Anti-CSF1R Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Leverantör: US Biological
USBI034268-APCEA 9900 SEK
USBI034268-APC
Anti-CSF1R Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50; Immunofluorescence: 1:10-50
Do not freeze!

Type: Primary
Antigen: CSF1R (colony stimulating factor 1 receptor)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR