Anti-CSF2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverantör: US Biological
USBI034270-HRPEA 9900 SEK
USBI034270-HRP
Anti-CSF2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50; Immunofluorescence: 1:10-50

Type: Primary
Antigen: CSF2 (colony stimulating factor 2 (granulocyte-macrophage))
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR