Anti-CSF2RA Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Leverantör: US Biological
USBI034271-PEEA 9900 SEK
USBI034271-PE
Anti-CSF2RA Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: CSF2RA (colony stimulating factor 2 receptor, alpha, low-affinity (granulocyte-macrophage))
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human