Anti-CSH1 Rabbit Polyclonal Antibody (Biotin)

Leverantör: US Biological
USBI034277-BIOTINEA 9900 SEK
USBI034277-BIOTIN
Anti-CSH1 Rabbit Polyclonal Antibody (Biotin)
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: CSH1 (chorionic somatomammotropin hormone 1 (placental lactogen))
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR