Anti-CSH1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Leverantör: US Biological
USBI034278-APCEA 9900 SEK
USBI034278-APC
Anti-CSH1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: CSH1 (chorionic somatomammotropin hormone 1 (placental lactogen))
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human