Anti-CSN1S1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Leverantör: United States Biological
USBI034282-PEEA 9900 SEK
USBI034282-PE
Anti-CSN1S1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: CSN1S1 (casein alpha s1)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human