Anti-CSNK1E Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverantör: United States Biological
USBI034286-FITCEA 9900 SEK
USBI034286-FITC
Anti-CSNK1E Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: CSNK1E (casein kinase 1, epsilon)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR