Anti-CSNK1G3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverantör: United States Biological
USBI034288-HRPEA 9900 SEK
USBI034288-HRP
Anti-CSNK1G3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Type: Primary
Antigen: CSNK1G3 (casein kinase 1, gamma 3)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse