Anti-CSRNP2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Leverantör: United States Biological
USBI034293-APEA 9900 SEK
USBI034293-AP
Anti-CSRNP2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: CSRNP2 (cysteine-serine-rich nuclear protein 2)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human