Anti-CSRNP3 Rabbit Polyclonal Antibody

Leverantör: United States Biological
USBI034295EA 6825 SEK
USBI034295
Anti-CSRNP3 Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: CSRNP3 (cysteine-serine-rich nuclear protein 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR