Anti-CSTB Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Leverantör: United States Biological
USBI034306-PEEA 9900 SEK
USBI034306-PE
Anti-CSTB Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: CSTB (cystatin B (stefin B))
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR