Anti-CSTF3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverantör: United States Biological
USBI034307-HRPEA 9900 SEK
USBI034307-HRP
Anti-CSTF3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: CSTF3 (cleavage stimulation factor, 3' pre-RNA, subunit 3, 77kDa)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR