Anti-FAM46D Rabbit Polyclonal Antibody (Biotin)
Leverantör: United States Biological
USBI034310-BIOTINEA 9900 SEK
USBI034310-BIOTIN
Anti-FAM46D Rabbit Polyclonal Antibody (Biotin)
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Type: Primary
Antigen: FAM46D (family with sequence similarity 46, member D)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human