Anti-ANKRD45 Rabbit Polyclonal Antibody

Leverantör: United States Biological
USBI034311EA 6825 SEK
USBI034311
Anti-ANKRD45 Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: ANKRD45 (ankyrin repeat domain 45)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR