Anti-CT173 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Leverantör: United States Biological
USBI034312-FITCEA 9900 SEK
USBI034312-FITC
Anti-CT173 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!
Type: Primary
Antigen: CT173
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human