Anti-CTDSPL2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverantör: United States Biological
USBI034326-FITCEA 9900 SEK
USBI034326-FITC
Anti-CTDSPL2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: CTDSPL2 (CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase like 2)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR