Anti-CTGF Rabbit Polyclonal Antibody

Leverantör: United States Biological
USBI034330EA 6825 SEK
USBI034330
Anti-CTGF Rabbit Polyclonal Antibody
Antikroppar
  • Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: CTGF (connective tissue growth factor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR