Anti-CTNNB1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Leverantör: United States Biological
USBI034336-PEEA 9900 SEK
USBI034336-PE
Anti-CTNNB1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: CTNNB1 (catenin (cadherin-associated protein), beta 1, 88kDa)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR