Anti-CTNND2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Leverantör: United States Biological
USBI034339-FITCEA 9900 SEK
USBI034339-FITC
Anti-CTNND2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: CTNND2 (catenin (cadherin-associated protein), delta 2)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse