Anti-CTRL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Leverantör: United States Biological
USBI034344-FITCEA 9900 SEK
USBI034344-FITC
Anti-CTRL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: CTRL (chymotrypsin-like)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human