Anti-CUL4B Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Leverantör: United States Biological
USBI034367-APEA 9900 SEK
USBI034367-AP
Anti-CUL4B Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: CUL4B (cullin 4B)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human