Anti-CUZD1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Leverantör: US Biological
USBI034372-APCEA 9900 SEK
USBI034372-APC
Anti-CUZD1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50
Do not freeze!
Type: Primary
Antigen: CUZD1 (CUB and zona pellucida-like domains 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human