Anti-CWC22 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Leverantör: US Biological
USBI034374-PEEA 9900 SEK
USBI034374-PE
Anti-CWC22 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: CWC22 (CWC22 spliceosome-associated protein homolog (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR