Anti-CXCL11 Rabbit Polyclonal Antibody

Leverantör: US Biological
USBI034380EA 6825 SEK
USBI034380
Anti-CXCL11 Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: CXCL11 (chemokine (C-X-C motif) ligand 11)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR