Anti-CXCR5 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Leverantör: US Biological
USBI034386-APEA 9900 SEK
USBI034386-AP
Anti-CXCR5 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: CXCR5 (chemokine (C-X-C motif) receptor 5)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human