Anti-CXCR6 Rabbit Polyclonal Antibody

Leverantör: US Biological
USBI034387EA 6825 SEK
USBI034387
Anti-CXCR6 Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: CXCR6 (chemokine (C-X-C motif) receptor 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR