Anti-CYB5A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverantör: US Biological
USBI034403-FITCEA 9900 SEK
USBI034403-FITC
Anti-CYB5A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: CYB5A (cytochrome b5 type A (microsomal))
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR