Anti-CYB5A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverantör: US Biological
USBI034403-HRPEA 9900 SEK
USBI034403-HRP
Anti-CYB5A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Type: Primary
Antigen: CYB5A (cytochrome b5 type A (microsomal))
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human