Anti-CYP11B2 Rabbit Polyclonal Antibody

USBI034422EA 5700 SEK
USBI034422
Anti-CYP11B2 Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:10-1:50; Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: CYP11B2 (cytochrome P450, family 11, subfamily B, polypeptide 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR