Anti-CYP2C19 Rabbit Polyclonal Antibody

Leverantör: US Biological
USBI034433EA 6825 SEK
USBI034433
Anti-CYP2C19 Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-100; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50

Type: Primary
Antigen: CYP2C19 (cytochrome P450, family 2, subfamily C, polypeptide 19)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR