Anti-MT-CYB Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverantör: US Biological
USBI034444-HRPEA 9900 SEK
USBI034444-HRP
Anti-MT-CYB Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: MT-CYB (mitochondrially encoded cytochrome b)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human