Anti-D4S234E Rabbit Polyclonal Antibody (Biotin)

Leverantör: US Biological
USBI034452-BIOTINEA 9900 SEK
USBI034452-BIOTIN
Anti-D4S234E Rabbit Polyclonal Antibody (Biotin)
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: D4S234E
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR