Anti-DACH1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverantör: US Biological
USBI034456-APEA 9900 SEK
USBI034456-AP
Anti-DACH1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: DACH1 (dachshund family transcription factor 1)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR