Vill du logga in igen?
Anti-DACH2 Rabbit Polyclonal Antibody (Biotin)
Leverantör: US Biological
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: DACH2 (dachshund family transcription factor 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
ORDER
Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
200 µl 200 µl USBI034457-BIOTIN
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFIKATIONER
Antigen symbol DACH2
Antigen name dachshund family transcription factor 2
Reaktivitet Human
Conjugation Biotin
Clonality Polyclonal
Antikroppstyp Primary
Värd Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Aminosyretal 144 to 173
Epitop N-terminal
Rening purified by Protein A chromatography
Form liquid
Storage buffer PBS, pH 7,2
Förvaringstemperatur Cool
Shipping temperature Ice
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal